Assay Details
Gene Symbol: LOC100523684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021062994.1 | XP_020918653.1 | 229 | 76 | ||
XM_021063008.1 | XP_020918667.1 | 208 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021062994.1, XM_021063008.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
