Assay Details
| Gene Symbol: CNIH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353763.5 | XP_003353811.1 | 641 | 107 | ||
| XM_003353764.4 | XP_003353812.1 | 332 | 107 | |||
| XM_021082810.1 | XP_020938469.1 | 715 | 107 | |||
More Information
Set Membership: |
|

CNIH2
Genomic Map