Assay Details
Gene Symbol: DNAJB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001924319.4 | XP_001924354.1 | 444 | 75 | ||
XM_005671091.2 | XP_005671148.1 | 444 | 75 | |||
XM_005671092.1 | XP_005671149.1 | 444 | 75 | |||
XM_013983329.1 | XP_013838783.1 | 444 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
