Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021089114.1 | XP_020944773.1 | 518 | 69 | ||
XM_021089115.1 | XP_020944774.1 | 581 | 69 | |||
XM_021089116.1 | XP_020944775.1 | 523 | 69 | |||
XM_021089117.1 | XP_020944776.1 | 515 | 69 | |||
XM_021089118.1 | XP_020944777.1 | 457 | 69 | |||
XM_021089119.1 | XP_020944778.1 | 520 | 69 | |||
XM_021089120.1 | XP_020944779.1 | 462 | 69 | |||
XM_021089121.1 | XP_020944780.1 | 330 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
