Assay Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021079288.1 | XP_020934947.1 | 1021 | 82 | ||
XM_021079289.1 | XP_020934948.1 | 1013 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
