Assay Details
Gene Symbol: GEMIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131811.5 | XP_003131859.3 | 269 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003131811.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
