Assay Details
| Gene Symbol: MINPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927672.4 | XP_001927707.1 | 1123 | 74 | ||
| XM_021072285.1 | XP_020927944.1 | 875 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MINPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927672.4 | XP_001927707.1 | 1123 | 74 | ||
| XM_021072285.1 | XP_020927944.1 | 875 | 74 | |||
Set Membership: |
|