Assay Details
| Gene Symbol: GPR35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075949.1 | XP_020931608.1 | 533 | 85 | ||
| XM_021075952.1 | XP_020931611.1 | 732 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021075949.1, XM_021075952.1 transcripts.
Set Membership: |
|

GPR35
Genomic Map