Assay Details
| Gene Symbol: FAM171A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066865.1 | XP_020922524.1 | 946 | 54 | ||
| XM_021066866.1 | XP_020922525.1 | 1031 | 54 | |||
| XM_021066867.1 | XP_020922526.1 | 449 | 54 | |||
More Information
Set Membership: |
|

FAM171A2
Genomic Map