Assay Details
| Gene Symbol: C2H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994676.2 | XP_013850130.1 | 497 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2H19orf57| Gene Symbol: C2H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013994676.2 | XP_013850130.1 | 497 | 87 | ||