Assay Details
| Gene Symbol: C2H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661242.3 | XP_005661299.1 | 1916 | 68 | ||
| XM_005661243.3 | XP_005661300.1 | 2118 | 68 | |||
| XM_013994675.2 | XP_013850129.1 | 1940 | 68 | |||
| XM_013994676.2 | XP_013850130.1 | 1835 | 68 | |||
| XM_013994679.2 | XP_013850133.1 | 1780 | 68 | |||
| XM_021083707.1 | XP_020939366.1 | 2888 | 68 | |||
More Information
Set Membership: |
|

C2H19orf57
Genomic Map