Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124463.6 | XP_003124511.4 | 870 | 114 | ||
| XM_021086364.1 | XP_020942023.1 | 974 | 114 | |||
More Information
Set Membership: |
|

ZNF713
Genomic Map