Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124463.6 | XP_003124511.4 | 355 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003124463.6 transcripts.
Set Membership: |
|

ZNF713
Genomic Map