Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670987.3 | XP_005671044.3 | 528 | 77 | ||
| XM_021072373.1 | XP_020928032.1 | 471 | 77 | |||
| XM_021072374.1 | XP_020928033.1 | 806 | 77 | |||
| XM_021072376.1 | XP_020928035.1 | 359 | 77 | |||
More Information
Set Membership: |
|

ZNF248
Genomic Map