Assay Details
| Gene Symbol: HSD3B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124487.4 | XP_003124535.2 | 1877 | 64 | ||
| XM_005655085.3 | XP_005655142.1 | 1894 | 64 | |||
| XM_021086410.1 | XP_020942069.1 | 1789 | 64 | |||
More Information
Set Membership: |
|

HSD3B7
Genomic Map