Assay Details
| Gene Symbol: BICRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003356632.5 | XP_003356680.2 | 2573 | 68 | ||
| XM_021098728.1 | XP_020954387.1 | 3329 | 68 | |||
| XM_021098729.1 | XP_020954388.1 | 3390 | 68 | |||
| XM_021098730.1 | XP_020954389.1 | 2404 | 68 | |||
| XM_021098731.1 | XP_020954390.1 | 2343 | 68 | |||

BICRAL
Genomic Map