Assay Details
| Gene Symbol: C12H17orf50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067409.1 | XP_020923068.1 | 140 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021067409.1 transcripts.
Set Membership: |
|

C12H17orf50
Genomic Map