Assay Details
Gene Symbol: POU6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081722.1 | XP_020937381.1 | 612 | 73 | ||
XM_021081723.1 | XP_020937382.1 | 406 | 73 | |||
XM_021081724.1 | XP_020937383.1 | 613 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
