Assay Details
| Gene Symbol: HEXIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005668721.3 | XP_005668778.2 | 801 | 106 | ||
| XM_013980886.2 | XP_013836340.1 | 709 | 106 | |||
| XM_013980887.2 | XP_013836341.1 | 712 | 106 | |||
| XM_013980888.2 | XP_013836342.1 | 804 | 106 | |||
| XM_013980889.2 | XP_013836343.1 | 4929 | 106 | |||
| XM_013980891.2 | XP_013836345.1 | 5195 | 106 | |||
| XM_013980892.2 | XP_013836346.1 | 5198 | 106 | |||
| XM_013980893.2 | XP_013836347.1 | 386 | 106 | |||
| XM_013980894.2 | XP_013836348.1 | 498 | 106 | |||
| XM_021066851.1 | XP_020922510.1 | 207 | 106 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005668721.3, XM_013980886.2, XM_013980887.2, XM_013980888.2, XM_013980889.2, XM_013980891.2, XM_013980892.2, XM_013980893.2, XM_013980894.2, XM_021066851.1 transcripts.
Set Membership: |
|

HEXIM2
Genomic Map