Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123042.4 | XP_003123090.1 | 674 | 64 | ||
| XM_005654707.3 | XP_005654764.1 | 484 | 64 | |||
| XM_013987307.2 | XP_013842761.1 | 707 | 64 | |||
| XM_021084122.1 | XP_020939781.1 | 1058 | 64 | |||
More Information
Set Membership: |
|

DOHH
Genomic Map