Assay Details
Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021066506.1 | XP_020922165.1 | 406 | 91 | ||
XM_021066507.1 | XP_020922166.1 | 266 | 91 | |||
XM_021066508.1 | XP_020922167.1 | 209 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021066507.1, XM_021066508.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
