Assay Details
Gene Symbol: BLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003359055.4 | XP_003359103.1 | 2002 | 66 | ||
XM_013990301.2 | XP_013845755.1 | 3736 | 66 | |||
XM_021072720.1 | XP_020928379.1 | 1244 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
