Assay Details
| Gene Symbol: GPR85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985693.2 | XP_013841147.1 | 1068 | 113 | ||
| XM_013985694.2 | XP_013841148.1 | 1522 | 113 | |||
| XM_013985695.2 | XP_013841149.1 | 1065 | 113 | |||
| XM_013985696.2 | XP_013841150.1 | 1092 | 113 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013985693.2, XM_013985694.2, XM_013985695.2, XM_013985696.2 transcripts.
Set Membership: |
|

GPR85
Genomic Map