Assay Details
| Gene Symbol: C15H2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013982266.2 | XP_013837720.1 | 235 | 102 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H2orf66| Gene Symbol: C15H2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013982266.2 | XP_013837720.1 | 235 | 102 | ||
Set Membership: | 
		   
					
						
							 |