Assay Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924273.7 | XP_001924308.5 | 369 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924273.7 | XP_001924308.5 | 369 | 72 | ||