Assay Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086295.1 | XP_020941954.1 | 912 | 102 | ||
| GenBank mRNA | AK236133.1 | - | 9 - 10 | 863 | 102 | |
| AK347758.1 | - | 9 - 10 | 827 | 102 | ||
| AK391667.1 | - | 9 - 10 | 859 | 102 | ||
More Information
Set Membership: |
|

TMEM120A
Genomic Map