Assay Details
Gene Symbol: GADD45GIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003123339.4 | XP_003123387.1 | 391 | 115 | ||
GenBank mRNA | AK346636.1 | - | 1 - 2 | 358 | 115 | |
AK399696.1 | - | 1 - 2 | 365 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
