Assay Details
Gene Symbol: C1GALT1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003135326.5 | XP_003135374.2 | 176 | 102 | ||
GenBank mRNA | AK391282.1 | - | 1 - 2 | 95 | 102 | |
AY610012.1 | - | 1 - 2 | 126 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135326.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
