Assay Details
| Gene Symbol: MRPL36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672623.2 | XP_005672680.1 | 668 | 73 | ||
| GenBank mRNA | AK233680.1 | - | 2 | 520 | 73 | |
| AK392116.1 | - | 2 | 514 | 73 | ||
More Information
Set Membership: |
|

MRPL36
Genomic Map