Assay Details
| Gene Symbol: ACTR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928877.5 | XP_001928912.1 | 2575 | 78 | ||
| GenBank mRNA | AK237460.1 | - | 1 | 1320 | 78 | |
| AK389443.1 | - | 5 | 1750 | 78 | ||
| AK390851.1 | - | 11 | 2518 | 78 | ||
More Information
Set Membership: |
|

ACTR1A
Genomic Map