Assay Details
| Gene Symbol: BICD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088132.1 | XP_020943791.1 | 6502 | 63 | ||
| XM_021088133.1 | XP_020943792.1 | 4598 | 63 | |||
| GenBank mRNA | AK239139.1 | - | 1 | 1119 | 63 | |
More Information
Set Membership: |
|

BICD2
Genomic Map