Assay Details
| Gene Symbol: GLRX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928993.6 | XP_001929028.1 | 64 | |||
| GenBank mRNA | AK389549.1 | - | 2 | 618 | 64 | |
| AY609918.1 | - | 2 | 64 | |||
More Information
Set Membership: |
|

GLRX5
Genomic Map