Assay Details
| Gene Symbol: PCMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988075.2 | XP_013843529.1 | 2191 | 116 | ||
| XM_021089409.1 | XP_020945068.1 | 1979 | 116 | |||
| XM_021089410.1 | XP_020945069.1 | 2393 | 116 | |||
| XM_021089411.1 | XP_020945070.1 | 1991 | 116 | |||
| XM_021089412.1 | XP_020945071.1 | 1511 | 116 | |||
| GenBank mRNA | AK389837.1 | - | 3 | 1123 | 116 | |
More Information
Set Membership: |
|

PCMTD1
Genomic Map