Assay Details
| Gene Symbol: DMRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094461.1 | XP_020950120.1 | 1631 | 90 | ||
| XM_021094462.1 | XP_020950121.1 | 2108 | 90 | |||
More Information
Set Membership: |
|

DMRTC2
Genomic Map