Assay Details
| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244460.1 | NP_001231389.1 | 3 - 4 | 480 | 89 | |
| XM_005658930.3 | XP_005658987.1 | 1561 | 89 | |||
| XM_005658931.3 | XP_005658988.1 | 1560 | 89 | |||
| XM_005658932.3 | XP_005658989.1 | 1554 | 89 | |||
| GenBank mRNA | AK352370.1 | - | 3 - 4 | 480 | 89 | |
More Information
Set Membership: |
|

SLC39A2
Genomic Map