Assay Details
| Gene Symbol: RPL36AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213917.1 | NP_999082.1 | 1 - 2 | 35 | 100 | |
| GenBank mRNA | AB000910.1 | - | 1 - 2 | 35 | 100 | |
| AK392526.1 | - | 1 - 2 | 54 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_213917.1 transcripts.
Set Membership: |
|

RPL36AL
Genomic Map