Assay Details
| Gene Symbol: RTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134358.4 | NP_001127830.4 | 1 | 3723 | 71 | |
| XM_021098142.1 | XP_020953801.1 | 3869 | 71 | |||
| GenBank mRNA | EU781029.4 | - | 1 | 3723 | 71 | |
More Information
Set Membership: |
|

RTL1
Genomic Map