Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167637.1 | NP_001161109.1 | 1 - 2 | 139 | 90 | |
XM_021098157.1 | XP_020953816.1 | 962 | 90 | |||
XM_021098158.1 | XP_020953817.1 | 701 | 90 | |||
GenBank mRNA | AB527053.1 | - | 1 - 2 | 139 | 90 | |
AK234930.1 | - | 2 - 3 | 921 | 90 | ||
AK396081.1 | - | 2 - 3 | 523 | 90 | ||
AK400421.1 | - | 2 - 3 | 281 | 90 | ||
AK400679.1 | - | 2 - 3 | 576 | 90 | ||
AY609698.1 | - | 2 - 3 | 216 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
