Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167637.1 | NP_001161109.1 | 3 - 4 | 346 | 76 | |
XM_021098157.1 | XP_020953816.1 | 1169 | 76 | |||
XM_021098158.1 | XP_020953817.1 | 908 | 76 | |||
GenBank mRNA | AB527053.1 | - | 3 - 4 | 346 | 76 | |
AK232554.1 | - | 4 - 5 | 780 | 76 | ||
AK234296.1 | - | 3 - 4 | 971 | 76 | ||
AK234930.1 | - | 4 - 5 | 1128 | 76 | ||
AK396081.1 | - | 4 - 5 | 730 | 76 | ||
AK400421.1 | - | 4 - 5 | 488 | 76 | ||
AK400679.1 | - | 4 - 5 | 783 | 76 | ||
AY609698.1 | - | 4 - 5 | 423 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
