Assay Details
Gene Symbol: COA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243594.1 | NP_001230523.1 | 2 - 3 | 318 | 82 | |
GenBank mRNA | AK233958.1 | - | 2 - 3 | 318 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
