Assay Details
| Gene Symbol: CES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243625.1 | NP_001230554.1 | 9 - 10 | 1193 | 102 | |
| GenBank mRNA | AK232431.1 | - | 9 - 10 | 1193 | 102 | |
| AK393090.1 | - | 9 - 10 | 1195 | 102 | ||
More Information
Set Membership: |
|

CES3
Genomic Map