Assay Details
| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243671.1 | NP_001230600.1 | 4 - 5 | 339 | 99 | |
| GenBank mRNA | AK240590.1 | - | 4 - 5 | 339 | 99 | |
More Information
Set Membership: |
|

RBMX2
Genomic Map