Assay Details
| Gene Symbol: DPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243616.1 | NP_001230545.1 | 3 | 1916 | 111 | |
| GenBank mRNA | EW616542.2 | - | 1 | 155 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243616.1 | NP_001230545.1 | 3 | 1916 | 111 | |
| GenBank mRNA | EW616542.2 | - | 1 | 155 | 111 | |
Set Membership: |
|