Assay Details
Gene Symbol: skp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY316293.1 | - | 751 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
