Assay Details
Gene Symbol: NAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY625683.1 | - | 949 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
