Assay Details
Gene Symbol: LTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF302788.1 | - | 212 | 77 | ||
AY796184.1 | - | 152 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF302788.1 | - | 212 | 77 | ||
AY796184.1 | - | 152 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() |