Assay Details
| Gene Symbol: AGL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AM502882.1 | - | 652 | 62 | ||
| DQ512360.1 | - | 690 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL14| Gene Symbol: AGL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AM502882.1 | - | 652 | 62 | ||
| DQ512360.1 | - | 690 | 62 | |||
Set Membership: |
|