Assay Details
| Gene Symbol: LOC100136976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU293126.1 | - | 452 | 86 | ||
| KJ551527.1 | - | 482 | 86 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100136976| Gene Symbol: LOC100136976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU293126.1 | - | 452 | 86 | ||
| KJ551527.1 | - | 482 | 86 | |||
Set Membership: |
|