Assay Details
Gene Symbol: MADS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF265562.1 | - | 6 - 7 | 710 | 87 | |
AY256861.1 | - | 5 - 6 | 290 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
