Assay Details
Gene Symbol: MLO15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU360796.1 | - | 13 - 14 | 1515 | 97 | |
EU812236.1 | - | 13 - 14 | 1515 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
